4-(2-Bromoethyl)tetrahydropyran 97%

Supplier: Ambeed
Danger

A247105-100MG A247105-1G A247105-250MG A247105-5G
77288-922EA 30 USD
77288-922 77288-924 77288-926 77288-928
4-(2-Bromoethyl)tetrahydropyran 97%
4-(2-Bromoethyl)tetrahydropyran

Precaución: for Research Use Only

Formula: C₇H₁₃BrO
MW: 193.0800 g/mol
MDL Number: MFCD09800414
CAS Number: 4677-20-7
UN: 1760
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR