2-Amino-7-bromoquinazoline 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
7-Bromoquinazolin-2-amine
A247097-100MG
A247097-1G
A247097-250MG
A247097-5G
77261-932EA
11
USD
77261-932
77261-934
77261-936
77746-381
2-Amino-7-bromoquinazoline 97%
2-Amino-7-bromoquinazoline
Precaución: for Research Use Only
Formula:
C₈H₆BrN₃ |
MDL Number:
MFCD09027698 CAS Number: 190274-15-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...