2,6-Dimethyl-4-nitroanisole 96%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-1,3-dimethyl-5-nitrobenzene

A246915-1G A246915-250MG A246915-5G
77249-890EA 52.53 USD
77249-890 77249-892 77249-894
2,6-Dimethyl-4-nitroanisole 96%
2,6-Dimethyl-4-nitroanisole

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
MDL Number: MFCD00024537
CAS Number: 14804-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR