ω-Bromo-4-cyanoacetophenone 95%

Supplier: Ambeed
Danger

Synonyms: 4-(2-Bromoacetyl)benzonitrile, 4-(Bromoacetyl)benzonitrile, 4-(2-Bromoacetyl)benzenecarbonitrile, 2-Bromo-4'-cyanoacetophenone, Bromomethyl p-cyanophenyl ketone, 2-Bromo-4′-cyanoacetophenone, 4-Cyanophenacyl bromide, α-Bromo-4-cyanoacetophenone

A246914-100G A246914-25G A246914-5G A246914-10G A246914-1G
77261-926EA 255 USD
77261-926 77261-928 77261-930 77616-022 77748-446
ω-Bromo-4-cyanoacetophenone 95%
ω-Bromo-4-cyanoacetophenone

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MDL Number: MFCD00052931
CAS Number: 20099-89-2
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR