3,9-Bis(octadecyloxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane 98% Carbon 64.8-69.5 %
Supplier: Ambeed
|
Warning
|
A246913-1000G
A246913-100G
A246913-25G
A246913-500G
77276-200EA
210.12
USD
77276-200
77276-202
77276-204
77276-206
3,9-Bis(octadecyloxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane 98% Carbon 64.8-69.5 %
3,9-Bis(octadecyloxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane
Precaución: for Research Use Only
Formula:
C₄₁H₈₂O₆P₂ MW: 733.03 g/mol |
MDL Number:
MFCD00071522 CAS Number: 3806-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...