4,5-Diamino-1-(2-hydroxyethyl)pyrazole sulfate 98%

Supplier: Ambeed
Warning

A246695-100G A246695-25G A246695-5G
77288-910EA 40.17 USD
77288-910 77288-912 77288-914
4,5-Diamino-1-(2-hydroxyethyl)pyrazole sulfate 98%
4,5-Diamino-1-(2-hydroxyethyl)pyrazole sulfate

Precaución: for Research Use Only

Formula: C₅H₁₂N₄O₅S
MW: 240.2376 g/mol
MDL Number: MFCD07368638
CAS Number: 155601-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR