4-Ethyl-2-methoxyphenol 98%

Supplier: Ambeed
Warning

A246685-100G A246685-10G A246685-25G A246685-500G A246685-5G
77288-898EA 44 USD
77288-898 77288-900 77288-902 77288-904 77288-906 77288-908
4-Ethyl-2-methoxyphenol 98%
4-Ethyl-2-methoxyphenol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MDL Number: MFCD00038714
CAS Number: 2785-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR