4-Amino-6-hydroxypyrimidine 98%

Supplier: Ambeed
Warning

A246502-10G A246502-1G A246502-250MG A246502-25G A246502-5G
77288-884EA 67 USD
77288-884 77288-886 77288-888 77288-890 77288-892
4-Amino-6-hydroxypyrimidine 98%
4-Amino-6-hydroxypyrimidine

Precaución: for Research Use Only

Formula: C₄H₅N₃O
MDL Number: MFCD00051502
CAS Number: 1193-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR