2-Bromo-3-chloro-5-hydroxypyridine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-5-chloropyridin-3-ol

A246277-100MG A246277-1G A246277-250MG
77261-916EA 63.86 USD
77261-916 77261-918 77261-920
2-Bromo-3-chloro-5-hydroxypyridine 97%
2-Bromo-3-chloro-5-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₃BrClNO
MW: 208.4400 g/mol
MDL Number: MFCD09835244
CAS Number: 130284-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR