2-Iodobenzamide 98%

Supplier: Ambeed
Warning

A245876-100G A245876-10G A245876-1G A245876-250MG A245876-5G
77261-888EA 133.9 USD
77261-888 77261-890 77261-892 77261-894 77261-896
2-Iodobenzamide 98%
2-Iodobenzamide

Precaución: for Research Use Only

Formula: C₇H₆INO
MW: 247.04 g/mol
MDL Number: MFCD00099248
CAS Number: 3930-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR