2-Amino-5-chloropyrimidin-4(3H)-one 97%

Supplier: Ambeed
Warning

A245853-100MG A245853-250MG A245853-1G
77261-886EA 67.98 USD
77261-886 77616-018 77748-443
2-Amino-5-chloropyrimidin-4(3H)-one 97%
2-Amino-5-chloropyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₄H₄ClN₃O
MW: 145.54 g/mol
MDL Number: MFCD28962202
CAS Number: 89033-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR