(2S,3R,4R,5S,6R)-2-(3-(Benzo[b]thiophen-2-ylmethyl)-4-fluorophenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol (S)-Pyrrolidine-2-carboxylic acid ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ipragliflozin (L-Proline)
A245799-10MG
A245799-1MG
A245799-50MG
A245799-5MG
77288-876EA
11.33
USD
77288-876
77288-878
77288-880
77288-882
(2S,3R,4R,5S,6R)-2-(3-(Benzo[b]thiophen-2-ylmethyl)-4-fluorophenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol (S)-Pyrrolidine-2-carboxylic acid ≥98%
(2S,3R,4R,5S,6R)-2-(3-(Benzo[b]thiophen-2-ylmethyl)-4-fluorophenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol (S)-Pyrrolidine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₂₆H₃₀FNO₇S MW: 519.58 g/mol |
MDL Number:
MFCD27956926 CAS Number: 951382-34-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...