Magnesiumbis(trifluoromethylsulfonyl)imide 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
3-nitro-1H-pyrimidine-2,4-dione, Magnesium(II) triflimide, Bis(trifluoromethanesulphonyl)imide Magnesium(II) salt, 2,4(1H,3H)-Pyrimidinedione, nitro-, Magnesium bis(trifluoromethylsulphonyl)imide, Magnesium(II) Bis(trifluoromethanesulphonyl)imide, 3-Nitropyrimidine-2,4(1H,3H)-dione
A245766-1G
A245766-25G
A245766-5G
77249-870EA
12
USD
77249-870
77249-872
77249-874
Magnesiumbis(trifluoromethylsulfonyl)imide 98%
Magnesiumbis(trifluoromethylsulfonyl)imide
Precaución: for Research Use Only
Formula:
C₄F₁₂MgN₂O₈S₄ MW: 584.6 g/mol |
MDL Number:
MFCD14156020 CAS Number: 133395-16-1 UN: 1759 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...