4-Nitrobenzenesulfonyl Fluoride 97%

Supplier: Ambeed
Danger

Synonyms: 4-Nitrobenzene-1-sulphonyl fluoride

A245596-1G A245596-25G A245596-5G A245596-10G
77288-856EA 11 USD
77288-856 77288-858 77288-860 77616-014
4-Nitrobenzenesulfonyl Fluoride 97%
4-Nitrobenzenesulfonyl Fluoride

Precaución: for Research Use Only

Formula: C₆H₄FNO₄S
MW: 205.16 g/mol
MDL Number: MFCD00009637
CAS Number: 349-96-2
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR