2-Bromo-3,5-dichloroisonicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-3,5-dichloropyridine-4-carboxylic acid

A245555-1G A245555-250MG A245555-5G
77261-880EA 28.84 USD
77261-880 77261-882 77261-884
2-Bromo-3,5-dichloroisonicotinic acid 98%
2-Bromo-3,5-dichloroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂NO₂
MW: 270.90 g/mol
MDL Number: MFCD03411637
CAS Number: 343781-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR