4-((6-Aminopyrimidin-4-yl)amino)benzenesulfonamide hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: PNU112455A HCl

A245488-10MG A245488-1MG A245488-50MG A245488-5MG
77238-176EA 57.68 USD
77238-176 77238-178 77238-180 77238-182
4-((6-Aminopyrimidin-4-yl)amino)benzenesulfonamide hydrochloride 97%
4-((6-Aminopyrimidin-4-yl)amino)benzenesulfonamide hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClN₅O₂S
MW: 301.75 g/mol
MDL Number: MFCD00023272
CAS Number: 21886-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR