7-Bromo-5-fluoro-3,4-dihydronaphthalen-1(2H)-one 97%
Supplier: Ambeed
|
Warning
|
A245416-100MG
A245416-250MG
A245416-1G
77261-876EA
33
USD
77261-876
77261-878
77568-386
7-Bromo-5-fluoro-3,4-dihydronaphthalen-1(2H)-one 97%
7-Bromo-5-fluoro-3,4-dihydronaphthalen-1(2H)-one
Precaución: For research use only.
Formula:
C₁₀H₈BrFO |
MDL Number:
MFCD18207828 CAS Number: 925442-95-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...