2-Mercapto-4-methylpyrimidine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methylpyrimidine-2(1H)-thione hydrochloride

A245377-100G A245377-1G A245377-25G A245377-500G A245377-5G
77261-866EA 118.45 USD
77261-866 77261-868 77261-870 77261-872 77261-874
2-Mercapto-4-methylpyrimidine hydrochloride 98%
2-Mercapto-4-methylpyrimidine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₇ClN₂S
MW: 162.635 g/mol
MDL Number: MFCD00012783
CAS Number: 6959-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR