4'-Benzyloxy-3'-methoxyacetophenone 97%

Supplier: Ambeed
Warning

A245343-10G A245343-1G A245343-25G
77276-162EA 42 USD
77276-162 77276-164 77593-562
4'-Benzyloxy-3'-methoxyacetophenone 97%
4'-Benzyloxy-3'-methoxyacetophenone

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₃
MDL Number: MFCD00017326
CAS Number: 1835-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR