Methyl 3-amino-2,6-dichloroisonicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-2,6-dichloropyridine-4-carboxylate

A245282-250MG A245282-25G A245282-5G
77276-156EA 44.29 USD
77276-156 77276-158 77276-160
Methyl 3-amino-2,6-dichloroisonicotinate 97%
Methyl 3-amino-2,6-dichloroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₆Cl₂N₂O₂
MW: 221.041 g/mol
MDL Number: MFCD07437844
CAS Number: 883107-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR