2-Phenoxypyridine 98%

Supplier: Ambeed
Warning

A245238-10G A245238-1G A245238-250MG
77276-150EA 141.11 USD
77276-150 77276-152 77276-154
2-Phenoxypyridine 98%
2-Phenoxypyridine

Precaución: for Research Use Only

Formula: C₁₁H₉NO
MW: 171.2 g/mol
MDL Number: MFCD00047432
CAS Number: 4783-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR