Methyl-3-amino-5-nitrobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-5-nitrobenzoate

A245181-10G A245181-1G A245181-250MG A245181-5G
77276-142EA 167.89 USD
77276-142 77276-144 77276-146 77276-148
Methyl-3-amino-5-nitrobenzoate 97%
Methyl 3-amino-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₈N₂O₄
MW: 196.1600 g/mol
MDL Number: MFCD00461253
CAS Number: 23218-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR