5-Aminoindole 98%

Supplier: Ambeed
Warning

Synonyms: Indol-5-ylamine, 5-Indolamine, 5-Amino-1H-indole, 1H-Indol-5-amine

A245121-100MG A245121-10G A245121-1G A245121-250MG A245121-25G A245121-5G
77249-856EA 5 USD
77249-856 77249-858 77249-860 77249-862 77249-864 77249-866
5-Aminoindole 98%
5-Aminoindole

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 190
MDL Number: MFCD00005679
CAS Number: 5192-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR