2-Methyl-5-nitroindole 96%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-5-nitro-1H-indole

A245067-10G A245067-1G A245067-25G A245067-5G
77261-856EA 154.5 USD
77261-856 77261-858 77261-860 77261-862
2-Methyl-5-nitroindole 96%
2-Methyl-5-nitroindole

Precaución: for Research Use Only

Formula: C₉H₈N₂O₂
MW: 176.17 g/mol
MDL Number: MFCD00090335
CAS Number: 7570-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR