5-Chloro-1-methyl-4-nitroimidazole 97%

Supplier: Ambeed
Warning

A245055-100G A245055-10G A245055-5G A245055-1000G A245055-25G A245055-500G
77276-128EA 51 USD
77276-128 77276-130 77276-132 77748-434 77748-435 77748-436
5-Chloro-1-methyl-4-nitroimidazole 97%
5-Chloro-1-methyl-4-nitroimidazole

Precaución: for Research Use Only

Formula: C₄H₄ClN₃O₂
MDL Number: MFCD00233664
CAS Number: 4897-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR