3-Methoxy-2-nitrobenzaldehyde 97%

Supplier: Ambeed
Warning

A245001-10G A245001-1G A245001-250MG A245001-5G
77238-168EA 253 USD
77238-168 77238-170 77238-172 77238-174
3-Methoxy-2-nitrobenzaldehyde 97%
3-Methoxy-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MDL Number: MFCD00007135
CAS Number: 53055-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR