2,3,5-Trifluoroaniline 97%

Supplier: Ambeed
Danger

A244999-10G A244999-1G A244999-5G A244999-25G
77288-826EA 98.88 USD
77288-826 77288-828 77288-830 77616-012
2,3,5-Trifluoroaniline 97%
2,3,5-Trifluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄F₃N
MW: 147.1 g/mol
MDL Number: MFCD00083549
CAS Number: 363-80-4
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR