4-Amino-1-isoindolinone 95%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2,3-dihydro-1H-isoindol-1-one

A244949-100MG A244949-1G A244949-250MG A244949-5G A244949-10G
77288-818EA 46.35 USD
77288-818 77288-820 77288-822 77288-824 77748-433
4-Amino-1-isoindolinone 95%
4-Amino-1-isoindolinone

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.17 g/mol
MDL Number: MFCD08234883
CAS Number: 366452-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR