2-(Methylthio)benzo[d]oxazole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(Methylthio)benzoxazole
A244947-100G
A244947-1G
A244947-25G
A244947-5G
77276-120EA
389.34
USD
77276-120
77276-122
77276-124
77276-126
2-(Methylthio)benzo[d]oxazole 98%
2-(Methylthio)benzo[d]oxazole
Precaución: for Research Use Only
Formula:
C₈H₇NOS MW: 165.2100 g/mol |
MDL Number:
MFCD00160027 CAS Number: 13673-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...