4-Chloro-6-methoxy-1H-indole 95%

Supplier: Ambeed
Warning

A244595-100MG A244595-1G A244595-250MG
77288-812EA 73.13 USD
77288-812 77288-814 77288-816
4-Chloro-6-methoxy-1H-indole 95%
4-Chloro-6-methoxy-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈ClNO
MW: 181.619 g/mol
MDL Number: MFCD00238559
CAS Number: 93490-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR