tert-Butyl 6-aminohexanoate 97%

Supplier: Ambeed
Warning

A244589-100MG A244589-1G A244589-250MG A244589-5G
77261-850EA 51.5 USD
77261-850 77261-852 77261-854 77550-570
tert-Butyl 6-aminohexanoate 97%
tert-Butyl 6-aminohexanoate

Precaución: for Research Use Only

Formula: C₁₀H₂₁NO₂
MW: 187.27 g/mol
MDL Number: MFCD12804788
CAS Number: 5514-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR