4-((4-((4-Aminophenyl)sulfonyl)phenyl)amino)-4-oxobutanoic acid 97%

Supplier: Ambeed
Warning

A244444-100MG A244444-1G A244444-250MG
77288-802EA 100 USD
77288-802 77288-804 77288-806
4-((4-((4-Aminophenyl)sulfonyl)phenyl)amino)-4-oxobutanoic acid 97%
4-((4-((4-Aminophenyl)sulfonyl)phenyl)amino)-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂O₅S
MW: 348.37 g/mol
MDL Number: MFCD00864826
CAS Number: 5934-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR