N-(4-((3,5-Difluorophenyl)sulfonyl)benzyl)imidazo[1,2-a]pyridine-6-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: GNE-617

A244376-10MG A244376-1MG A244376-2MG A244376-5MG
77238-144EA 106.09 USD
77238-144 77238-146 77238-148 77238-150
N-(4-((3,5-Difluorophenyl)sulfonyl)benzyl)imidazo[1,2-a]pyridine-6-carboxamide 98%
N-(4-((3,5-Difluorophenyl)sulfonyl)benzyl)imidazo[1,2-a]pyridine-6-carboxamide

Precaución: for Research Use Only

Formula: C₂₁H₁₅F₂N₃O₃S
MW: 427.42 g/mol
MDL Number: MFCD26960964
CAS Number: 1362154-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR