(2R,3R,4S,5R,6S)-2-((Benzoyloxy)methyl)-6-(ethylthio)-4-hydroxytetrahydro-2H-pyran-3,5-diyl dibenzoate 97%
Supplier: Ambeed
|
Warning
|
A244193-10G
A244193-1G
77261-846EA
74.16
USD
77261-846
77261-848
(2R,3R,4S,5R,6S)-2-((Benzoyloxy)methyl)-6-(ethylthio)-4-hydroxytetrahydro-2H-pyran-3,5-diyl dibenzoate 97%
(2R,3R,4S,5R,6S)-2-((Benzoyloxy)methyl)-6-(ethylthio)-4-hydroxytetrahydro-2H-pyran-3,5-diyl dibenzoate
Precaución: for Research Use Only
Formula:
C₂₉H₂₈O₈S MW: 536.59 g/mol |
MDL Number:
MFCD18643040 CAS Number: 1423018-01-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...