Ethyl 2-(4-methylpent-2-enoyl)thiazole-4-carboxylate 97%

Supplier: Ambeed
Warning

A243595-10G A243595-250MG
77249-828EA 280.16 USD
77249-828 77249-830
Ethyl 2-(4-methylpent-2-enoyl)thiazole-4-carboxylate 97%
Ethyl 2-(4-methylpent-2-enoyl)thiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₃S
MW: 253.32 g/mol
MDL Number: MFCD27635175
CAS Number: 944559-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR