tert-Butyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydroquinoline-1(2H)-carboxylate 97%

Supplier: Ambeed
Warning

A243569-100MG A243569-1G A243569-250MG A243569-5G
77276-094EA 35.02 USD
77276-094 77276-096 77276-098 77568-362
tert-Butyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydroquinoline-1(2H)-carboxylate 97%
tert-Butyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydroquinoline-1(2H)-carboxylate

Precaución: For research use only.

Formula: C₂₀H₃₀BNO₄
MW: 359.26 g/mol
MDL Number: MFCD11858571
CAS Number: 1912446-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR