4-Methylphthalimide 98%

Supplier: Ambeed
Warning

A243144-1G A243144-25G A243144-5G
77288-776EA 26 USD
77288-776 77288-778 77288-780
4-Methylphthalimide 98%
4-Methylphthalimide

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MDL Number: MFCD05155224
CAS Number: 40314-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR