4'-Bromo-3'-fluoroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-3-fluoro--acetophenone

A243078-100MG A243078-10G A243078-1G A243078-250MG A243078-5G A243078-100G
77276-074EA 5.15 USD
77276-074 77276-076 77276-078 77276-080 77276-082 77748-430
4'-Bromo-3'-fluoroacetophenone 98%
4'-Bromo-3'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆BrFO
MW: 217.04 g/mol
MDL Number: MFCD00466241
CAS Number: 304445-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR