3-(Cyclopropylmethoxy)-4-(difluoromethoxy)benzaldehyde 97%

Supplier: Ambeed
Warning

A243077-100G A243077-1G A243077-250MG A243077-25G A243077-5G A243077-10G
77288-766EA 1662.42 USD
77288-766 77288-768 77288-770 77288-772 77288-774 77748-429
3-(Cyclopropylmethoxy)-4-(difluoromethoxy)benzaldehyde 97%
3-(Cyclopropylmethoxy)-4-(difluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₂F₂O₃
MW: 242.22 g/mol
MDL Number: MFCD07779381
CAS Number: 151103-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR