4-Methoxy-N,N-diphenylaniline 97%

Supplier: Ambeed
Warning

A243073-10G A243073-1G A243073-250MG A243073-25G A243073-5G
77288-756EA 112 USD
77288-756 77288-758 77288-760 77288-762 77288-764
4-Methoxy-N,N-diphenylaniline 97%
4-Methoxy-N,N-diphenylaniline

Precaución: for Research Use Only

Formula: C₁₉H₁₇NO
MDL Number: MFCD03929020
CAS Number: 4316-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR