trans,trans-4'-Pentyl-4-bicyclohexylmethanol 97%

Supplier: Ambeed
Warning

Synonyms: ((trans,trans)-4'-Pentyl-[1,1'-bi(cyclohexan)]-4-yl)methanol, trans,trans-4'-Amyl-4-bicyclohexylmethanol

A242970-100G A242970-1G A242970-25G A242970-5G
77238-134EA 270.89 USD
77238-134 77238-136 77238-138 77238-140
trans,trans-4'-Pentyl-4-bicyclohexylmethanol 97%
trans,trans-4'-Pentyl-4-bicyclohexylmethanol

Precaución: for Research Use Only

Formula: C₁₈H₃₄O
MW: 266.47 g/mol
MDL Number: MFCD16036281
CAS Number: 82598-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR