3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride 98%

Supplier: Ambeed
Warning

Synonyms: 3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride

A242426-1000G A242426-100G A242426-10G A242426-25G A242426-500G A242426-5G
77276-044EA 331 USD
77276-044 77276-046 77276-048 77276-050 77276-052 77276-054
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride 98%
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride

Precaución: for Research Use Only

Formula: C₁₃H₁₆ClNOS
MDL Number: MFCD00011959
CAS Number: 4568-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR