3-Hydroxy-2-nitropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Nitropyridin-3-ol, 2-Nitro-3-pyridinol

A242403-100G A242403-10G A242403-25G A242403-500G A242403-5G
77276-034EA 53 USD
77276-034 77276-036 77276-038 77276-040 77276-042
3-Hydroxy-2-nitropyridine 98%
3-Hydroxy-2-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₄N₂O₃
MDL Number: MFCD00006258
CAS Number: 15128-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR