2-Chloro-4,6-dimethylnicotinonitrile 97%
Supplier: Ambeed
|
Danger
|
A242188-10G
A242188-1G
A242188-250MG
A242188-25G
A242188-5G
77238-118EA
19
USD
77238-118
77238-120
77238-122
77238-124
77238-126
2-Chloro-4,6-dimethylnicotinonitrile 97%
2-Chloro-4,6-dimethylnicotinonitrile
Precaución: for Research Use Only
Formula:
C₈H₇ClN₂ |
MDL Number:
MFCD00051676 CAS Number: 14237-71-9 UN: 3439 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...