N2,N4-Diallyl-6-(4-(bis(4-fluorophenyl)methyl)piperazin-1-yl)-1,3,5-triazine-2,4-diamine dimethanesulfonate 97%

Supplier: Ambeed
Warning

Synonyms: Almitrine Bismesylate

A242169-100MG A242169-1G A242169-5G A242169-10G A242169-250MG A242169-25G
77249-810EA 5.15 USD
77249-810 77249-812 77615-998 77746-365 77746-366 77746-367
N2,N4-Diallyl-6-(4-(bis(4-fluorophenyl)methyl)piperazin-1-yl)-1,3,5-triazine-2,4-diamine dimethanesulfonate 97%
N2,N4-Diallyl-6-(4-(bis(4-fluorophenyl)methyl)piperazin-1-yl)-1,3,5-triazine-2,4-diamine dimethanesulfonate

Precaución: for Research Use Only

Formula: C₂₈H₃₇F₂N₇O₆S₂
MW: 669.76 g/mol
MDL Number: MFCD01679060
CAS Number: 29608-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR