Methyl 4-bromo-2-hydroxy-6-methylbenzoate 97%

Supplier: Ambeed
Warning

A242061-100MG A242061-250MG A242061-1G
77261-810EA 175 USD
77261-810 77261-812 77615-996
Methyl 4-bromo-2-hydroxy-6-methylbenzoate 97%
Methyl 4-bromo-2-hydroxy-6-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MDL Number: MFCD30528900
CAS Number: 2089319-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR