7-Chloro-2-(furan-2-yl)-[1,2,4]triazolo[1,5-c]pyrimidin-5-amine 97%

Supplier: Ambeed
Warning

A242014-100MG A242014-1G A242014-250MG
77249-804EA 66.95 USD
77249-804 77249-806 77249-808
7-Chloro-2-(furan-2-yl)-[1,2,4]triazolo[1,5-c]pyrimidin-5-amine 97%
7-Chloro-2-(furan-2-yl)-[1,2,4]triazolo[1,5-c]pyrimidin-5-amine

Precaución: for Research Use Only

Formula: C₉H₆ClN₅O
MW: 235.62 g/mol
MDL Number: MFCD26391812
CAS Number: 213896-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR