2,6-Bis(2-benzimidazolyl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Bis(benzimidazol-2-yl)pyridine, 2,2'-(2,6-Pyridinediyl)bis(benzimidazole)

A241983-1G A241983-250MG A241983-5G A241983-25G
77249-798EA 39 USD
77249-798 77249-800 77249-802 77746-363
2,6-Bis(2-benzimidazolyl)pyridine 98%
2,6-Bis(2-benzimidazolyl)pyridine

Precaución: for Research Use Only

Formula: C₁₉H₁₃N₅
MDL Number: MFCD00134500
CAS Number: 28020-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR