(5-Methoxy-3-methylpyridin-2-yl)methanamine 97%

Supplier: Ambeed
Warning

A241891-100MG A241891-1G A241891-250MG
77238-112EA 336.81 USD
77238-112 77238-114 77238-116
(5-Methoxy-3-methylpyridin-2-yl)methanamine 97%
(5-Methoxy-3-methylpyridin-2-yl)methanamine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O
MW: 152.19 g/mol
MDL Number: MFCD18255406
CAS Number: 1256792-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR