4-(4,5-Diphenyl-1H-imidazol-2-yl)benzoic acid 98%

Supplier: Ambeed
Warning

A241840-1G A241840-5G
77288-746EA 18.54 USD
77288-746 77288-748
4-(4,5-Diphenyl-1H-imidazol-2-yl)benzoic acid 98%
4-(4,5-Diphenyl-1H-imidazol-2-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₂₂H₁₆N₂O₂
MW: 340.38 g/mol
MDL Number: MFCD00999654
CAS Number: 5496-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR